Anti-ANOS1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-11053R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Anosmin 1
- Antigen symbol:ANOS1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Adhesion molecule-like X-linked|Kallmann syndrome protein|KALIG1|KAL|ADMLX|KALM|KALIG 1|Kallmann syndrome interval gene 1|KALM_HUMAN|HHA|Kallmann syndrome 1 sequence anosmin 1|KMS.|KAL1|Anosmin-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
May be an adhesion-like molecule with anti-protease activity.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ANOS1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat