Order Entry
Sweden
ContactUsLinkComponent
 
Anti-RPSA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-11015R-A350
 :  Bioss
Anti-RPSA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-11015R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    Ribosomal Protein SA
  • Antigen symbol:
    RPSA
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    3921
  • Antigen synonyms:
    LRP/LR|SA|37LRP|LBP/p40|67LR|LAMBR|ICAS|lamR|LAMR1|p40|NEM/1CHD4|LRP|LBP
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

Phakinin is a membrane-associated and cytoskeletal intermediate filament (IF) protein specific to the eye lens. IFs are cytoskeletal structures that typically contain a head, rod and tail domain. Unlike most IFs, Phakinin completely lacks the C-terminal tail domain thus contributing to the unique structure of the beaded filament that is specific to the lens. Phakinin is required for the assembly of beaded filaments and cytoskeletal networks that are important for the long-term maintenance of optical properties and transparency of the lens. Phakinin copolymerizes with Filensin, another IF protein, to form the 10-nm filamentous structures of the beaded filaments. Phakinin is also capable of self-assembling into filament-like structures that form thicker bundles. Mutations in the gene encoding Phakinin can result in lens cataract.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RPSA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat