Order Entry
Sweden
ContactUsLinkComponent
Anti-OPCML Rabbit Polyclonal Antibody (Cy5®)
Anti-OPCML Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1081R-CY5
Supplier:  Bioss
Anti-OPCML Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-1081R-CY5
Supplier:  Bioss

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    Opioid binding Protein/Cell Adhesion molecule-like
  • Antigensymbol:
    OPCML
  • Klonalitet:
    Polyclonal
  • Konjugation:
    Cy5®
  • Värd:
    Rabbit
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG
  • Reaktivitet:
    Human,
    Rat,
    Mouse
  • Korsad adsorption:
    No
  • Form:
    liquid
  • Gen-ID:
    4978
  • Antigen synonymer:
    IGLON1|OBCAM|OPCM
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Förvaringstemperatur:
    Store at 4°C for 12 months
  • Koncentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Rening:
    Purified by Protein A
  • Förp.:
    100 µl

Specifications

About this item

This gene encodes a member of the IgLON subfamily in the immunoglobulin protein superfamily. The encoded protein is localized in the plasma membrane and may have an accessory role in opioid receptor function. This gene has an ortholog in rat and bovine. The opioid binding-cell adhesion molecule encoded by the rat gene binds opioid alkaloids in the presence of acidic lipids, exhibits selectivity for mu ligands and acts as a GPI-anchored protein. Since the encoded protein is highly conserved in species during evolution, it may have a fundamental role in mammalian systems. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OPCML
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat