Anti-NRF2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-1074R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Nuclear Respiratory Factor 2
- Antigen symbol:NRF2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:4780
- Antigen synonyms:NFE2-related factor 2|nuclear factor|HEBP1|erythroid derived 2|NRF2|nuclear factor erythroid-derived 2-like 2|nuclear factor erythroid 2-related factor 2|NF-E2-related factor 2|like 2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:430-480/605
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Transcription activator that binds to antioxidant response (ARE) elements in the promoter regions of target genes. Important for the coordinated up-regulation of genes in response to oxidative stress. May be involved in the transcriptional activation of genes of the beta-globin cluster by mediating enhancer activity of hypersensitive site 2 of the beta-globin locus control region.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Nrf2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat