Anti-PTGFR Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Catalog # BOSSBS-10433R-A488
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Prostaglandin F Receptor
- Antigen symbol:PTGFR
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 488
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5737
- Antigen synonyms:SMAP-6|EWI-F|FPRP|CD9P-1|CD315
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:176-198/359
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Receptor for prostaglandin F2-alpha (PGF2-alpha). The activity of this receptor is mediated by G proteins which activate a phosphatidylinositol-calcium second messenger system. Initiates luteolysis in the corpus luteum (By similarity). Isoforms 2 to 7 do not bind PGF2-alpha but are proposed to modulate signaling by participating in variant receptor complexes; heterodimers between isoform 1 and isoform 5 are proposed to be a receptor for prostamides including the synthetic analog bimatoprost.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PTGFR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat