Specifikationer
- Antibody type:Primary
- Antigen name:HGV Polyprotein
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifikationer
Om denna produkt
Hepatitis G virus is from a group of viruses called GB viruses (GB virus A, GB virus B and GB virus C) and is also designated as GB virus C (GBV C). HGV is a positive stranded RNA virus, with a genome of 10 KB and appears, on the basis of genome length and organisation, to be a member of the Flaviviridae and distinct from the hepatitis C virus (HCV). HGV infection appears to be transmitted following receipt of blood from HGV RNA positive donors, however, these patients have only moderate liver enzyme elevations and do not develop chronic liver disease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HGV Polyprotein
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus