Anti-E1 glycoprotein Rabbit Polyclonal Antibody (Cy7®)
BOSSBS-0319R-CY7
: Bioss
- Antibody type:Primary
- Antigen name:E1 glycoprotein
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Virus
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: E1 glycoprotein
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus