Anti-CRP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-0155R-A555
: Bioss
- Antibody type:Primary
- Antigen name:C-Reactive Protein
- Antigen symbol:CRP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1401
- Antigen synonyms:CRP1|MGC149895|C-reactive protein|pentraxin 1|CRP|D1S181E|RP11-419N10.4|CSRP|CYRP|MGC88244|PTX1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:140-190/224
- Purification:Purified by Protein A
- Pk:100 µl
Displays several functions associated with host defense: it promotes agglutination, bacterial capsular swelling, phagocytosis and complement fixation through its calcium-dependent binding to phosphorylcholine. Can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CRP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity: