Glass tubings, CONTURAX®
Supplier: Schott AG
Certificates
About this item
Borosilicate glass tubing, varying types of inner and outer shapes.
- High chemical resistance
- High hydrolytic resistance
- High resistance to thermal shock
For glassblown laboratory ware, pressed glass articles (e.g. balls, lenses), technical articles, and other laboratory/industrial technology.