Anti-JunB Rabbit Polyclonal Antibody [clone: D253]
Catalog # BWRLBS1196
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:JunB
- Clonality:Polyclonal
- Clone:D253
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 220-270 of Human JunB.
JunB (D253) pAb detects endogenous levels of JunB protein.
Type: Primary
Antigen: JunB
Clonality: Polyclonal
Clone: D253
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat