Anti-Catenin-beta Rabbit Polyclonal Antibody [clone: I35]
BWRLBS1041
- Antibody type:Primary
- Antigen symbol:Catenin- beta
- Clonality:Polyclonal
- Clone:I35
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids N-terminus of Human Catenin-beta.
Catenin-? (I35) pAb detects endogenous levels of Catenin-? protein.
Type: Primary
Antigen: Catenin-beta
Clonality: Polyclonal
Clone: I35
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat