Anti-Cleaved-Lamin A Rabbit Polyclonal Antibody [clone: D230-]
BWRLBS7013
- Antibody type:Primary
- Antigen symbol:Cleaved-Lamin A
- Clonality:Polyclonal
- Clone:D230-
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 180-230 of Human Lamin A.
Cleaved-Lamin A (D230-) pAb detects the small fragment (28 kDa) of lamin A and lamin C resulting from cleavage at Asp230. The antibody does not cross-react with full length lamin A or lamin C.
Type: Primary
Antigen: Cleaved-Lamin A
Clonality: Polyclonal
Clone: D230-
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat