Anti-GADD 45gamma Rabbit Polyclonal Antibody [clone: S123]
BWRLBS9140
- Antibody type:Primary
- Antigen symbol:GADD 45 gamma
- Clonality:Polyclonal
- Clone:S123
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 101-150 of Human GADD 45gamma.
GADD 45? (S123) pAb detects endogenous levels of GADD 45? protein.
Type: Primary
Antigen: GADD 45gamma
Clonality: Polyclonal
Clone: S123
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat