Anti-PDYN Rabbit Polyclonal Antibody
Catalog # BWRLBS7886
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PDYN
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human PDYN.
PDYN pAb detects endogenous levels of PDYN protein.
Type: Primary
Antigen: PDYN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human