Anti-p-Rb Rabbit Polyclonal Antibody [clone: S807]
Catalog # BWRLBS4166
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-Rb
- Clonality:Polyclonal
- Clone:S807
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Rb around the phosphorylation site of Serine 807.
p-Rb (S807) pAb detects endogenous levels of Rb protein only when phosphorylated at Ser807
Type: Primary
Antigen: p-Rb
Clonality: Polyclonal
Clone: S807
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat