Anti-Trypsin-1 Rabbit Polyclonal Antibody
Catalog # BWRLBS6284
Leverantör: Bioworld Technology
Specifikationer
- Antikroppstyp:Primär
- Antigensymbol:Trypsin-1
- Klonalitet:Polyclonal
- Värd:Rabbit
- ImmunKemi:Yes
- Reaktivitet:Human,Rat,Mouse
- Western blot:Yes
- Storlek:100 µg
- Förp.:100 µG
Specifikationer
Om denna produkt
Recombinant full length Human Trypsin-1.
Trypsin-1 pAb detects endogenous levels of Trypsin-1 protein.
Type: Primary
Antigen: Trypsin-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat