Anti-p-ErbB2/HER2 Rabbit Polyclonal Antibody [clone: Y1221/Y1222]
BWRLBS4089
- Antibody type:Primary
- Antigen symbol:p-ErbB2/HER2
- Clonality:Polyclonal
- Clone:Y1221/Y1222
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human ErbB2/HER2 around the phosphorylation site of Tyrosine 1221/1222.
p-ErbB2/HER2 (Y1221/Y1222) pAb detects endogenous levels of ErbB2/HER2 protein only when phosphorylated at Tyr1221 and Tyr1222.
Type: Primary
Antigen: p-ErbB2/HER2
Clonality: Polyclonal
Clone: Y1221/Y1222
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat