Anti-PI3K p85beta Rabbit Polyclonal Antibody
Catalog # BWRLBS60314
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PI3K p85 beta
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A synthetic peptide corresponding to residues in Human PI3K p85beta.
PI 3-kinase p85? pAb detects endogenous levels of PI 3-kinase p85? protein.
Type: Primary
Antigen: PI3K p85beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat