Anti-eIF4E Rabbit Polyclonal Antibody [clone: R215]
100 µg
Catalog # BWRLBS5516
Supplier: Bioworld Technology
Anti-eIF4E Rabbit Polyclonal Antibody [clone: R215]
Catalog # BWRLBS5516
Supplier: Bioworld Technology
Jump to:
Product Details & Documents
Synthetic peptide, corresponding to amino acids 181-230 of Human eIF4E.
eIF4E (R215) pAb detects endogenous levels of eIF4E protein.
Type: Primary
Antigen: eIF4E
Clonality: Polyclonal
Clone: R215
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Western blot:Yes
- Cat. No.:BWRLBS5516
- Clonality:Polyclonal
- Reactivity:Human|Rat|Mouse
- Antibody type:Primary
- Host:Rabbit
- ImmunoChemistry:Yes
- Antigen symbol:eIF4E
- Clone:R215
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



