Anti-Dyskerin Rabbit Polyclonal Antibody
Catalog # BWRLBS6586
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Dyskerin
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human Dyskerin.
Dyskerin pAb detects endogenous levels of Dyskerin protein.
Type: Primary
Antigen: Dyskerin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat