Order Entry
Sweden
ContactUsLinkComponent
 
Anti-p21 Rabbit Polyclonal Antibody [clone: FL]
  BWRLBS6561
Anti-p21 Rabbit Polyclonal Antibody [clone: FL]
  BWRLBS6561

 

  • Antibody type:
    Primary
  • Antigen symbol:
    p21
  • Clonality:
    Polyclonal
  • Clone:
    FL
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recombinant full length Human P21.

p21 (FL) pAb detects endogenous levels of p21 protein.

Type: Primary
Antigen: p21
Clonality: Polyclonal
Clone: FL
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat