Anti-p-GRF-1 Rabbit Polyclonal Antibody [clone: Y1087]
Catalog # BWRLBS4814
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-GRF-1
- Clonality:Polyclonal
- Clone:Y1087
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human GRF-1 around the phosphorylation site of Tyrosine 1087.
p-GRF-1 (Y1087) pAb detects endogenous levels of GRF-1 protein only when phosphorylated at Tyr1087.
Type: Primary
Antigen: p-GRF-1
Clonality: Polyclonal
Clone: Y1087
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat