Order Entry
Sweden
ContactUsLinkComponent
 
Anti-p-GluR1 Rabbit Polyclonal Antibody [clone: S863]
  BWRLBS4793
Anti-p-GluR1 Rabbit Polyclonal Antibody [clone: S863]
  BWRLBS4793

 

  • Antibody type:
    Primary
  • Antigen symbol:
    p-GluR1
  • Clonality:
    Polyclonal
  • Clone:
    S863
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic phosphopeptide derived from human GluR1 around the phosphorylation site of Serine 863.

p-GluR1 (S863) pAb detects endogenous levels of GluR1 protein only when phosphorylated at Ser863.

Type: Primary
Antigen: p-GluR1
Clonality: Polyclonal
Clone: S863
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat