Anti-hnRNP E1 Rabbit Polyclonal Antibody
Catalog # BWRLBS60055
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:hnRNP E1
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
A synthetic peptide corresponding to residues in Human hnRNP E1.
hnRNP E1 pAb detects endogenous levels of hnRNP E1 protein.
Type: Primary
Antigen: hnRNP E1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat