Anti-TCP-1epsilon Rabbit Polyclonal Antibody
Catalog # BWRLBS5958
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:TCP-1 epsilon
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 295-340 of Human TCP-1epsilon.
TCP-1? pAb detects endogenous levels of TCP-1? protein.
Type: Primary
Antigen: TCP-1epsilon
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat