Anti-PTPRC Rabbit Polyclonal Antibody
Catalog # BWRLBS6900
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase receptor type C
- Antigen symbol:PTPRC
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant full length Human PTPRC
PTPRC pAb detects endogenous levels of PTPRC protein.
Type: Primary
Antigen: PTPRC
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human