Order Entry
Sweden
ContactUsLinkComponent
Anti-Catenin-beta Rabbit Polyclonal Antibody
Anti-Catenin-beta Rabbit Polyclonal Antibody
Catalog # BWRLBS6879
Anti-Catenin-beta Rabbit Polyclonal Antibody
Catalog # BWRLBS6879

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Catenin- beta
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recombinant full length Human Catenin-beta.

CTNNB1 pAb detects endogenous levels of CTNNB1 protein.

Type: Primary
Antigen: Catenin-beta
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat