Anti-p-CDC25A Rabbit Polyclonal Antibody [clone: S124]
Catalog # BWRLBS4668
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-CDC25A
- Clonality:Polyclonal
- Clone:S124
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human Cdc25A around the phosphorylation site of Serine 124.
p-CDC25A (S124) pAb detects endogenous levels of Cdc25A protein only when phosphorylated at Ser124.
Type: Primary
Antigen: p-CDC25A
Clonality: Polyclonal
Clone: S124
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat