Anti-p-CD19 Rabbit Polyclonal Antibody [clone: Y531]
Catalog # BWRLBS4664
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-CD19
- Clonality:Polyclonal
- Clone:Y531
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human CD19 around the phosphorylation site of Tyrosine 531.
p-CD19 (Y531) pAb detects endogenous levels of CD19 protein only when phosphorylated at Tyr531.
Type: Primary
Antigen: p-CD19
Clonality: Polyclonal
Clone: Y531
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat