Order Entry
Sweden
ContactUsLinkComponent
Anti-p-PPARgamma Rabbit Polyclonal Antibody [clone: S112]
Anti-p-PPARgamma Rabbit Polyclonal Antibody [clone: S112]
Catalog # BWRLBS4444
Anti-p-PPARgamma Rabbit Polyclonal Antibody [clone: S112]
Catalog # BWRLBS4444

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    p-PPAR gamma
  • Clonality:
    Polyclonal
  • Clone:
    S112
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic phosphopeptide derived from human PPARgamma around the phosphorylation site of Serine 112.

p-PPAR? (S112) pAb detects endogenous levels of PPAR? protein only when phosphorylated at Ser112.

Type: Primary
Antigen: p-PPARgamma
Clonality: Polyclonal
Clone: S112
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat