Anti-p-WAVE1 Rabbit Polyclonal Antibody [clone: Y125]
BWRLBS4370
- Antibody type:Primary
- Antigen symbol:p-WAVE1
- Clonality:Polyclonal
- Clone:Y125
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human WAVE1 around the phosphorylation site of Tyrosine 125.
p-WAVE1 (Y125) pAb detects endogenous levels of WAVE1 protein only when phosphorylated at Tyr125
Type: Primary
Antigen: p-WAVE1
Clonality: Polyclonal
Clone: Y125
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat