Anti-p-PKC-pan Rabbit Polyclonal Antibody [clone: alphaT497]
BWRLBS4348
- Antibody type:Primary
- Antigen symbol:p-PKC-pan
- Clonality:Polyclonal
- Clone:αT497
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human PKC-pan around the phosphorylation site of Threonine 497.
p-PKC-pan (?T497) pAb detects endogenous levels of PKC PKC ?, ? I, ? II, ?, ?, ?, ?, ? and ? isoforms only when phosphorylated at a residue homologous to threonine 497 of human PKC?.
Type: Primary
Antigen: p-PKC-pan
Clonality: Polyclonal
Clone: alphaT497
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat