Anti-ZNF225 Rabbit Polyclonal Antibody [clone: L667]
Catalog # BWRLBS2675
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:ZNF225
- Clonality:Polyclonal
- Clone:L667
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 634-682 of Human ZNF225.
ZNF225 (L667) pAb detects endogenous levels of ZNF225 protein.
Type: Primary
Antigen: ZNF225
Clonality: Polyclonal
Clone: L667
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human