Anti-EDG-5 Rabbit Polyclonal Antibody [clone: L303]
Catalog # BWRLBS2594
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:EDG-5
- Clonality:Polyclonal
- Clone:L303
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 281-330 of Human EDG-5.
EDG-5 (L303) pAb detects endogenous levels of EDG-5 protein.
Type: Primary
Antigen: EDG-5
Clonality: Polyclonal
Clone: L303
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat