Anti-PKD1/2/3 Rabbit Polyclonal Antibody [clone: I740]
Catalog # BWRLBS1685
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PKD1/2/3
- Clonality:Polyclonal
- Clone:I740
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 708-763 of Human PKD1.
PKD1/2/3 (I740) pAb detects endogenous levels of PKD1/2/3 protein.
Type: Primary
Antigen: PKD1/2/3
Clonality: Polyclonal
Clone: I740
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat