Anti-RPL34 Rabbit Polyclonal Antibody [clone: R76]
Catalog # BWRLBS3050
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:RPL34
- Clonality:Polyclonal
- Clone:R76
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 50-100 of Human Ribosomal Protein L34.
Ribosomal Protein L34 (R76) pAb detects endogenous levels of Ribosomal Protein L34 protein.
Type: Primary
Antigen: RPL34
Clonality: Polyclonal
Clone: R76
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat