Anti-GATA4 Rabbit Polyclonal Antibody [clone: Q258]
BWRLBS1647
- Antibody type:Primary
- Antigen symbol:GATA4
- Clonality:Polyclonal
- Clone:Q258
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 230-280 of Human GATA4.
GATA4 (Q258) pAb detects endogenous levels of GATA4 protein.
Type: Primary
Antigen: GATA4
Clonality: Polyclonal
Clone: Q258
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat