Anti-NY-CO-1 Rabbit Polyclonal Antibody [clone: K914]
Catalog # BWRLBS2456
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:NY-CO-1
- Clonality:Polyclonal
- Clone:K914
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 881-930 of Human NY-CO-1.
NY-CO-1 (K914) pAb detects endogenous levels of NY-CO-1 protein.
Type: Primary
Antigen: NY-CO-1
Clonality: Polyclonal
Clone: K914
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat