Anti-B-Myb Rabbit Polyclonal Antibody [clone: G573]
Catalog # BWRLBS1614
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:B-Myb
- Clonality:Polyclonal
- Clone:G573
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 543-590 of Human B-Myb.
B-Myb (G573/577) pAb detects endogenous levels of B-Myb protein.
Type: Primary
Antigen: B-Myb
Clonality: Polyclonal
Clone: G573
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat