Anti-Tuberin Rabbit Polyclonal Antibody [clone: F933]
Catalog # BWRLBS1604
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Tuberin
- Clonality:Polyclonal
- Clone:F933
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 900-950 of Human Tuberin.
Tuberin/TSC2 (F933) pAb detects endogenous levels of Tuberin/TSC2 protein.
Type: Primary
Antigen: Tuberin
Clonality: Polyclonal
Clone: F933
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat