Anti-ARA70 Rabbit Polyclonal Antibody [clone: L379 ]
BWRLBS2976
- Antibody type:Primary
- Antigen symbol:ARA70
- Clonality:Polyclonal
- Clone:L379
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 348-398 of Human ARA70.
ARA70 (L379) pAb detects endogenous levels of ARA70 protein
Type: Primary
Antigen: ARA70
Clonality: Polyclonal
Clone: L379
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat