Anti-GDF-9 Rabbit Polyclonal Antibody [clone: E322]
Catalog # BWRLBS2929
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:GDF-9
- Clonality:Polyclonal
- Clone:E322
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 301-350 of Human GDF-9.
GDF-9 (E322) pAb detects endogenous levels of GDF-9 protein.
Type: Primary
Antigen: GDF-9
Clonality: Polyclonal
Clone: E322
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human