Anti-SR-1E Rabbit Polyclonal Antibody [clone: A127]
BWRLBS2386
- Antibody type:Primary
- Antigen symbol:SR-1E
- Clonality:Polyclonal
- Clone:A127
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 95-140 of Human SR-1E.
SR-1E (A127) pAb detects endogenous levels of SR-1E protein.
Type: Primary
Antigen: SR-1E
Clonality: Polyclonal
Clone: A127
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human