Anti-HES-6 Rabbit Polyclonal Antibody [clone: A6]
Catalog # BWRLBS2344
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:HES-6
- Clonality:Polyclonal
- Clone:A6
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminal of Human HES-6.
HES-6 (A6) pAb detects endogenous levels of HES-6 protein.
Type: Primary
Antigen: HES-6
Clonality: Polyclonal
Clone: A6
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human