Anti-c-Src Rabbit Polyclonal Antibody [clone: S69]
Catalog # BWRLBS3754
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:c-Src
- Clonality:Polyclonal
- Clone:S69
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 40-80 of Human c-Src.
c-Src (S69) pAb detects endogenous levels of c-Src protein.
Type: Primary
Antigen: c-Src
Clonality: Polyclonal
Clone: S69
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat