Anti-hnRNP H Rabbit Polyclonal Antibody [clone: R192]
BWRLBS2286
- Antibody type:Primary
- Antigen symbol:hnRNP H
- Clonality:Polyclonal
- Clone:R192
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 180-230 of Human hnRNP H.
hnRNP H (R192) pAb detects endogenous levels of hnRNP H protein.
Type: Primary
Antigen: hnRNP H
Clonality: Polyclonal
Clone: R192
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat