Anti-CKMT2/sMtCK Rabbit Polyclonal Antibody [clone: W262]
Catalog # BWRLBS2260
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CKMT2/sMtCK
- Clonality:Polyclonal
- Clone:W262
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 230-280 of Human CKMT2.
sMtCK (W262) pAb detects endogenous levels of sMtCK protein.
Type: Primary
Antigen: CKMT2/sMtCK
Clonality: Polyclonal
Clone: W262
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat