Anti-Integrin beta1 Rabbit Polyclonal Antibody [clone: Y783]
Catalog # BWRLBS1444
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Integrin beta1
- Clonality:Polyclonal
- Clone:Y783
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 741-790 of Human Integrin beta1.
ITGB1 (Y783) pAb detects endogenous levels of ITGB1 protein.
Type: Primary
Antigen: Integrin beta1
Clonality: Polyclonal
Clone: Y783
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat