Anti-Trk B Rabbit Polyclonal Antibody [clone: V509]
Catalog # BWRLBS1431
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Trk B
- Clonality:Polyclonal
- Clone:V509
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 102-158 of Human Sarcoglycan-β.
Trk B (V509) pAb detects endogenous levels of Trk B protein.
Type: Primary
Antigen: Trk B
Clonality: Polyclonal
Clone: V509
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat