Anti-TBX22 Rabbit Polyclonal Antibody [clone: A30]
BWRLBS2173
- Antibody type:Primary
- Antigen symbol:TBX22
- Clonality:Polyclonal
- Clone:A30
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminual of Human TBX22.
TBX22 (A30) pAb detects endogenous levels of TBX22 protein.
Type: Primary
Antigen: TBX22
Clonality: Polyclonal
Clone: A30
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human